Skip to Content
Merck
All Photos(1)

Key Documents

AB15332

Sigma-Aldrich

Anti-Nicotinic Acetylcholine Receptor α7 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
rat, mouse, human
technique(s):
western blot: suitable
citations:
3

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Chrna7(11441)

Specificity

This rabbit polyclonal antibody specifically detects Nicotinic Receptor 7. It targets an epitope with 28 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 28 amino acids from the C-terminal half of human Nicotinic Receptor 7.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.Western Blotting Analysis: A 1:500 dilution of this antibody detected Nicotinic Acetylcholine Receptor 7 in Rat brain tissue lysate.


Target description

Neuronal acetylcholine receptor subunit alpha-7 (UniProt: P36544; also known as -Nicotinic Receptor Alpha 7, Nicotinic AChR alpha7) is encoded by the CHRNA7 (also known as NACHRA7) gene (Gene ID: 1139) in human. Nicotinic acetylcholine receptors are cation-selective, ligand-gated ion channels that mediate fast neurotransmission. They are distributed at the post-synaptic membrane of muscle cells. A similar neuronal-type ACh receptor is also found in the central nervous system and is involved in a wide range of physiological and pathological functions. They are assembled either as homo-pentamers of 7, 8, and 9 or hetero-pentamers of 2 6 in combination with β2 β4 or 9 with 10 subunits. They regulate the flow of sodium, potassium, and calcium ions across the cell membrane. The binding of ligands to these receptors triggers a tertiary conformational transition of neuronal AChRs among functionally distinct resting, open, and desensitized states. Nicotinic AChR 7 is a multi-pass, homopentameric, membrane glycoprotein that is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 23-230), four transmembrane domains (aa 231-255; 262-280; 296-317; and 470-490), and one cytoplasmic domain (aa 318-469). (Ref.: Ho, TN., et al. (2020). Front. Neurosci. 14; 609005; Unwin, N. (2013). Q. Rev. Biophys. 46(4); 283-322).

Linkage

Replaces: AB5637

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.02 M phosphate buffer, pH 7.6, 0.25 M NaCl, and 0.1% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Experimental and therapeutic medicine, 13(1), 85-90 (2017-01-27)
Endothelial progenitor cells (EPCs) have an important role in maintaining endothelial homeostasis. Previous studies reported that smoking has detrimental effects on EPCs; however, recent studies revealed that short-term nicotine exposure may benefit EPCs. As most smokers are exposed to nicotine
Brijesh K Garg et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 65(9), 499-512 (2017-08-02)
Alpha7 nicotinic acetylcholine receptors (α7 nAChRs) are important drug targets in neurological disorders and inflammation, making their detection and localization by validated antibodies highly desirable. However, tests in knockout animals raised questions about specificity of antibodies to mouse α7 nAChRs.
Arthur Antonio Ruiz Pereira et al.
PloS one, 19(4), e0299534-e0299534 (2024-04-04)
Alzheimer's disease (AD) is the most common neuronal disorder that leads to the development of dementia. Until nowadays, some therapies may alleviate the symptoms, but there is no pharmacological treatment. Microdosing lithium has been used to modify the pathological characteristics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service