Skip to Content
Merck
All Photos(1)

Key Documents

8.40006

Sigma-Aldrich

Tetradecyltrimethylammonium bromide

for synthesis

Synonym(s):

Tetradecyltrimethylammonium bromide, N,N,N-Trimethyl-1-tetradecanammonium bromide, Myristyltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H38BrN
CAS Number:
Molecular Weight:
336.39
MDL number:
UNSPSC Code:
12352107
EC Index Number:
214-291-9
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

powder

potency

3900 mg/kg LD50, oral (Rat)

mp

245-250 °C

solubility

100 g/L

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

[Br-].[N+](CCCCCCCCCCCCCC)(C)(C)C

InChI

1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CXRFDZFCGOPDTD-UHFFFAOYSA-M

Analysis Note

Assay (336,4 g/mole): ≥ 98.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service