Skip to Content
Merck
All Photos(1)

Key Documents

5.05986

Sigma-Aldrich

SR 95531

≥98% (HPLC), solid, GABAA receptor antagonist, Calbiochem®

Synonym(s):

SR95531 (Gabazine), GABA A Receptor Antagonist, SR-95531, Gabazine, SR95531, SR 95531, GABAA Receptor Antagonist, SR-95531, Gabazine, SR95531, SR 95531

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N3O3 · xHBr
CAS Number:
Molecular Weight:
287.31 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

SR95531 (Gabazine),

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

440 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 25 mM

storage temp.

2-8°C

SMILES string

[Br-].[N+H2]=C1N(N=C(C=C1)c2ccc(cc2)OC)CCCC(=O)O

InChI

1S/C15H17N3O3.BrH/c1-21-12-6-4-11(5-7-12)13-8-9-14(16)18(17-13)10-2-3-15(19)20;/h4-9,16H,2-3,10H2,1H3,(H,19,20);1H

InChI key

GFZHNFOGCMEYTA-UHFFFAOYSA-N

General description

A selective, competitive antagonist of GABAA receptor (IC50 = 440 nM) that does not affect pentobarbitone or etomidate- induced currents even at high concentrations (~100 µM). Also shown to block glycine receptors, but with low affinity (Kb = 190 µM).

Biochem/physiol Actions

Primary Target
GABAA

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Muroi, Y., et al. 2009. Biophys. J.96, 499.
Beato, M., et al. 2007. J. Physiol.580, 171.
Uchida, I., et al. 1996. Eur. J. Pharmacol.307, 89.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service