Skip to Content
Merck
All Photos(1)

Key Documents

495620

Sigma-Aldrich

Olomoucine

A purine derivative that acts as a potent, reversible and selective inhibitor of p34cdk1/cyclin B and related kinases.

Synonym(s):

Olomoucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N6O
CAS Number:
Molecular Weight:
298.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]1(c2nc(nc(c2nc1)NCc3ccccc3)NCCO)C

InChI

1S/C15H18N6O/c1-21-10-18-12-13(17-9-11-5-3-2-4-6-11)19-15(16-7-8-22)20-14(12)21/h2-6,10,22H,7-9H2,1H3,(H2,16,17,19,20)

InChI key

GTVPOLSIJWJJNY-UHFFFAOYSA-N

General description

A purine derivative that acts as a potent, reversible and selective inhibitor of p34cdk1/cyclin B (IC50 = 7 µM) and related kinases including p33cdk2/cyclin A (IC50 = 7 µM), p33cdk2/cyclin E (IC50 = 7 µM), p33cdk5/p35 (IC50 = 3 µM), and p44 MAP kinase (IC50 = 25 µM). Acts by competing for the ATP binding domain of the kinase. Exhibits reduced sensitivity towards related kinases, p34cdk4/cyclin D (IC50 >1 mM), and p40cdk6/cyclin D3 (IC50 >250 µM). Does not significantly affect the activity of other protein kinases at 1 mM. Known to inhibit DNA synthesis in interleukin-2-stimulated T lymphocytes and trigger G1 arrest similar to that observed in interleukin-2-deprived cells. Also used to synchronize cells in G1. A 50 mM (5 mg/336 µl) solution of Olomoucine (Cat. No. 495624) in DMSOis also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p34cdk1/cyclin B
Product competes with ATP.
Reversible: yes
Target IC50: 7 µM, 7 µM, 7 µM, 3 µM, 25 µM against p34cdk1/cyclin B, p33cdk2/cyclin A, p33cdk2/cyclin E, p33cdk5/p35, and p44 MAP kinase, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 6 months at -20°C.

Other Notes

Abraham, R.T., et al. 1995. Biol. Cell 83, 105.
Mistelli, T., and Warren, G. 1995. J. Cell Sci. 108, 2715.
Glab, N., et al. 1994. FEBS Lett. 353, 207.
Veselý, J., et al. 1994. Eur. J. Biochem.224, 771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of inhibitors in the study of MAPK signaling.
Yoav D Shaul et al.
Methods in molecular biology (Clifton, N.J.), 250, 113-126 (2004-02-03)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service