Skip to Content
Merck
All Photos(1)

Key Documents

419825

Sigma-Aldrich

Hh Signaling Antagonist X, Itraconazole

The Hh Signaling Antagonist X, Itraconazole, also referenced under CAS 84625-61-6, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Hh Signaling Antagonist X, Itraconazole, 14LDM Inhibitor, Oriconazole, R51211, Smo Antagonist VII, Sporanox, 14-α-demethylase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H38Cl2N8O4
CAS Number:
Molecular Weight:
705.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)Cl)[C@@]3(O[C@H](CO3)COc4ccc(cc4)N5CCN(CC5)c6ccc(cc6)[n]7[c]([n](nc7)C(CC)C)=O)C[n]2ncnc2

InChI

1S/C35H38Cl2N8O4/c1-3-25(2)45-34(46)44(24-40-45)29-7-5-27(6-8-29)41-14-16-42(17-15-41)28-9-11-30(12-10-28)47-19-31-20-48-35(49-31,21-43-23-38-22-39-43)32-13-4-26(36)18-33(32)37/h4-13,18,22-25,31H,3,14-17,19-21H2,1-2H3/t25?,31-,35-/m0/s1

InChI key

VHVPQPYKVGDNFY-ZPGVKDDISA-N

General description

A cell-permeable, orally available triazole-class antifungal agent that, in addition to its known inhibitory activity against fungal cytochrome P450 14-α-demethylase, also acts as an effective Hh signaling inhibitor (IC50 ~ 0.8 µM against SHHN-induced Gli transcription activity in Shh-Light2 cells) in a Ptch-independent manner (IC50 ~ 0.9 µM against Ptch promoter-mediated transcription activity in murine Ptch-/- fibroblasts), presumably by binding smoothened (smo) at a site distinct from that targeted by cyclopamine (Cat. No. 239803) and SAG (Cat. No. 566660), as Itraconazole does not compete with BODIPY-cyclopamine binding, and it alters only the magnitude, but not the EC50, of SAG-induced Gli transcription activity in Shh-Light2 cells. Shown to effectively suppress Hh pathway-dependent growth of both engrafted medulloblastoma and endogenous basal cell carcinoma in mice in vivo (37.5 to 100 mg/kg/12 hr, p.o.) in a reversible manner.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstituiton, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, J., et al. 2010. Cancer Cell17, 388.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service