Skip to Content
Merck
All Photos(1)

Key Documents

1.46316

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

closure type, with MPC connector and triple-bagged, bag of 1 × 10 L

Synonym(s):

TSB, CASO, Casein Soybean Digest Broth, Tryptic Soy Broth, Tryptone Soya Broth, SCDB, Tryptic Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP
USP

Quality Level

sterility

sterile; sterile-filtered

form

liquid (ready-to-use)

feature

closure type with MPC connector and triple-bagged
ready-to-use

packaging

bag of 1 × 10 L

pH

7.3 ( in H2O)

application(s)

aseptic process simulation
pharmaceutical

compatibility

stand alone

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Application: Aseptic process simulation, Media Fill Testing

Packaging: 10 L bag with MPC connector and triple-bagged

Synonym: TSB, CASO, Casein Soybean Digest Broth, Tryptic Soy Broth, Tryptone Soya Broth, SCDB
Tryptic Soy Broth is a universal complex medium for the isolation and cultivation of microorganisms, especially for the microbiological aseptic process simulation (Media Fill Test). The broth is suitable for media fill tests according to the recommendations of the “FDA Aseptic Guide” in aseptic production lines. The formulation of the basic medium (Soybean-Casein Digest Broth) is prepared according to the recommendations of the current European, and United States Pharmacopoeia.

Application

Tryptic Soy Broth is suitable for use in media fill tests. The included peptones and glucose allow the cultivation and isolation of a broad range of fastidious aerobic bacteria, yeasts and molds.

Other Notes

The ready-to-use and sterile medium is contained in a 10L self-collapsing bag, with an MPC connector.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

High quality granulated and ready-to-use culture media irradiated for accurate and reliable aseptic process simulations.

High quality granulated and ready-to-use culture media irradiated for accurate and reliable aseptic process simulations.

High quality granulated and ready-to-use culture media irradiated for accurate and reliable aseptic process simulations.

High quality granulated and ready-to-use culture media irradiated for accurate and reliable aseptic process simulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service