1.37007
Hydrochloric acid
fuming 37%, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, NF
Pharma Manufacturing
Synonym(s):
Hydrochloric acid, Hydrogen chloride solution
About This Item
Recommended Products
grade
GMO free
GMP
Halal
Kosher
Quality Level
Agency
BP
ChP
JP
NF
Ph. Eur.
vapor density
1.3 (vs air)
vapor pressure
3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)
product line
EMPROVE® EXPERT
technique(s)
API processing | salt formation: suitable
bp
>100 °C (lit.)
density
1.18 g/L at 20 °C
application(s)
cleaning in place
pharma/biopharma processes
foreign activity
Aflatoxin
storage temp.
2-25°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Packaging
1.37007.2500 / 2.5L / glass bottle
1.37007.9025 / 25L / PE canister
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service