Skip to Content
Merck
All Photos(2)

Key Documents

1.13350

Supelco

2-Propanol

≥99.8% (GC), for preparative chromatography Prepsolv

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-661-7
NACRES:
NB.21
Assay:
≥99.8% (GC)
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

2-Propanol, for preparative chromatography Prepsolv

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

750 °F

potency

5045 mg/kg LD50, oral (Rat)
12800 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-12.7 %, 93 °C

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤1.0 mg/L

transmittance

220 nm, ≥50%
260 nm, ≥98%

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

storage temp.

2-25°C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 1.0 mg/l
Water: ≤ 0.05 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 220 nm): ≥ 50 %
Transmission (from 260 nm): ≥ 98 %
Filtred by 0.2µm filter

Legal Information

PREPSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service