Skip to Content
Merck
All Photos(1)

Key Documents

1.06237

Supelco

N-(1-Naphthyl)ethylenediamine dihydrochloride

GR for analysis ACS

Synonym(s):

N-(1-Naphthyl)ethylenediamine dihydrochloride, Naphthylethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16Cl2N2
CAS Number:
Molecular Weight:
259.17
MDL number:
UNSPSC Code:
12352107
EC Index Number:
215-981-2
NACRES:
NA.05
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

form

solid

pH

1.0 (20 °C, 25 g/L in H2O)

solubility

30 g/L

bulk density

380 kg/m3

storage temp.

15-25°C

SMILES string

[Cl-].[Cl-].[N+H2](CC[N+H3])c1c2c(ccc1)cccc2

InChI

1S/C12H14N2.2ClH/c13-8-9-14-12-7-3-5-10-4-1-2-6-11(10)12;;/h1-7,14H,8-9,13H2;2*1H

InChI key

MZNYWPRCVDMOJG-UHFFFAOYSA-N

Linkage

Replaces: NX0230-1

Analysis Note

Assay (argentometric, calc. on anhydrous substance): ≥ 97.0 %
Identity (IR-spectrum): conforms
Appearance of solution (20 g/l, water): clear, dissolution is complete
1-Naphthylamine (HPLC): ≤ 0.1 %
2-Naphthylamine (HPLC): ≤ 0.01 %
Water (according to Karl Fischer): ≤ 5.0 %
Sensitivity to sulfanilamide: conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel González-Hedström et al.
Nutrients, 12(2) (2020-02-26)
Childhood obesity is associated with metabolic and cardiovascular comorbidities. The development of these alterations may have its origin in early life stages such as the lactation period through metabolic programming. Insulin resistance is a common complication in obese patients and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service