Skip to Content
Merck
All Photos(2)

Key Documents

07-1720

Sigma-Aldrich

Anti-FOXO4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fork head domain transcription factor AFX1, forkhead box O4, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
mouse, human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXO4(4303)
mouse ... Foxo4(54601)

General description

Forkhead box O4 (FOXO4 or AFX) is a member of the forkhead family of transcription factors, (FOXO), which play an important role in cellular proliferation, metabolism, stress tolerance, and aging. FOXO transcription factors have a characteristic forkhead domain and are located in the cytoplasm. FOXO4 has similar properties to FOXO1, FOXO3, and FOXO and is associated with acute leukemia. Upon phosphorylation, FOXO4 is translocated from the cytosol to the nucleus. FOXO4 has three phosphorylation sites at Thr32, Ser197, and Ser262. There are 2 pathways, Akt and reversible acetylation, involved in negative control of FOXO4, resulting in cell survival. FOXO4 represses smooth muscle cell (SMC) differentiation by interacting and suppressing myocardin. FOXO4 also down regulates HIF-1 alpha protein levels which prevent hypoxia response.

Specificity

This antibody recognizes FOXO4 at the N-terminus.

Immunogen

Epitope: N-terminus
Synthetic peptide corresponding to the N-terminal region of FOXO4.

Application

Anti-FOXO4 Antibody is an antibody against FOXO4 for use in WB & IC.
Immunocytochemistry:
A 1:500 dilution of this antibody was used to detect FOXO4 in NIH/3T3 and A431 cells. Positive nuclear staining.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Western Blot Analysis:
1:500 dilution of this antibody was used to detect FOXO4 in mouse kidney lysate.

Target description

~65 kDa

Linkage

Replaces: MABE539; MABD181

Physical form

Affinity purified
Purified rabbit polyclonal antibody precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.
To reconstitute the antibody, centrifuge the antibody at a moderate speed (5,000 rpm) for 5 minutes. Carefully remove the ammonium sulfate/PBS buffer solution and discard; 10μL of residual ammonium sulfate solution will not affect the re-suspension of the antibody. Do not let the protein pellet dry, as severe loss of antibody reactivity can occur. Re-suspend the antibody pellet in 100L either standard PBS or TBS (pH 7.3-7.5). DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 4-25°C prior to use. Small particles of precipitated antibody that fail to re-suspend are normal. Vials are overfilled to compensate for any losses.

Analysis Note

Control
Mouse kidney lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah M Senf et al.
American journal of physiology. Cell physiology, 300(6), C1490-C1501 (2011-03-11)
The Forkhead Box O (FOXO) transcription factors regulate diverse cellular processes, and in skeletal muscle are both necessary and sufficient for muscle atrophy. Although the regulation of FOXO by Akt is well evidenced in skeletal muscle, the current study demonstrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service