Skip to Content
Merck
All Photos(1)

Key Documents

T-020

Supelco

cis-Tramadol-13C, D3 hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC15D3H22NO2 · HCl
Molecular Weight:
303.85
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.N(C[C@@H]1[C@@](CCCC1)(O)c2cc(ccc2)OC([2H])([2H])[2H])(C)C

InChI

1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16+;/m1./s1/i3D3;

InChI key

PPKXEPBICJTCRU-QRPFLOSHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard for tramadol testing or isotope dilution methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing, pain prescription monitoring, or forensic analysis. Tramadol is a synthetic analgesic used to treat moderate to moderately severe pain. The drug, sold under trade names Ultram®, Ultracet®, Ralivia, and Tramal, has a wide range of applications from treatment of rheumatoid arthritis to fibromyalgia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Ultracet is a registered trademark of Johnson & Johnson
Ultram is a registered trademark of Johnson & Johnson

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article presents an LC-MS/MS method for the determination of tramadol in urine samples using reversed-phase LC-MS/MS aided by isotope internal standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service