Skip to Content
Merck
All Photos(1)

Key Documents

810241P

Avanti

TopFluor TMR PE

Avanti Research - A Croda Brand 810241P, powder

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3- phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H76 BF2N4 O10P
CAS Number:
Molecular Weight:
972.94
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810241P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810241P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoethanolamine contains a phosphate group linked with the second carbon of ethanolamine.

Biochem/physiol Actions

Phosphoethanolamine (PEA) is a precursor to phosphatidylethanolamine and phosphatidylcholine, involved in the biosynthesis of cell membrane.[1] In human body, PEA is a product of degradation of sphingosine phosphate and endocannabinoid anandamide.[2] PEA has potential cytotoxic properties and is known to induce the caspase-independent cell death. PEA is capable of inhibiting tumor growth in vitro and in vivo in mouse models.[3][1]

Packaging

5 mL Amber Glass Screw Cap Vial (810241P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisley I Mambelli et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 103, 18-28 (2018-04-11)
Phosphoethanolamine (PEA) is a fundamental precursor during the biosynthesis of cell membranes phospholipids. In the past few years, it has been described as a potential antitumor agent. In previous studies, we demonstrated that PEA showed antitumor properties in vitro and
Noam Pondé et al.
Ecancermedicalscience, 10, 681-681 (2016-12-03)
The use of unproven forms of therapy in cancer treatment is very common. In Brazil, the distribution by researchers to patients of an investigational agent called phophoethanolamine (PHOS) has led to a widely publicized scientific scandal. PHOS is a precursor
Dhaval Patel et al.
Oxidative medicine and cellular longevity, 2017, 4829180-4829180 (2017-08-09)
Phosphatidylethanolamine (PE) is the second most abundant phospholipid in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells; it is enriched in the inner leaflet of membranes, and it is especially abundant in the inner mitochondrial
Ann C Kimble-Hill et al.
The journal of physical chemistry. B, 122(35), 8404-8415 (2018-06-08)
Phosphatidylinositol (PI) lipids are necessary for many cellular signaling pathways of membrane associated proteins, such as angiomotin (Amot). The Amot family regulates cellular polarity, growth, and migration. Given the low concentration of PI lipids in these membranes, it is likely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service