Skip to Content
Merck
All Photos(1)

Key Documents

700099P

Avanti

lanosterol-95

Avanti Research - A Croda Brand

Synonym(s):

lanosterin; lanosta-8,24-dienol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

8,24-lanostadien-3β-ol

Assay

>95% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (700099P-100mg)
pkg of 1 × 5 mg (700099P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1C([C@H]2[C@](CC1)(C3=C([C@]4([C@@]([C@H](CC4)[C@@H](CCC=C(C)C)C)(CC3)C)C)CC2)C)(C)C

InChI

1S/C30H50O/c1-20(2)10-9-11-21(3)22-14-18-30(8)24-12-13-25-27(4,5)26(31)16-17-28(25,6)23(24)15-19-29(22,30)7/h10,21-22,25-26,31H,9,11-19H2,1-8H3/t21-,22-,25+,26+,28-,29-,30+/m1/s1

InChI key

CAHGCLMLTWQZNJ-BQNIITSRSA-N

General description

Lanosterol is a lanostane-type tetracyclic triterpene. It is the first sterol in lipid biosynthetic pathway.

Biochem/physiol Actions

Lanosterol blocks cataract formation, inhibits lens opacity and reverses crystalline aggregation.

Packaging

5 mL Amber Glass Screw Cap Vial (700099P-100mg)
5 mL Amber Glass Screw Cap Vial (700099P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetracyclic triterpenes from Inonotus obliquus (PERS.) PIL.(CHAGA) growing in Russia
Zhukovich EN, et al.
Pharmaceutical Chemistry Journal, 44(9), 495-496 (2010)
Lanosterol synthase pathway alleviates lens opacity in age-related cortical cataract
Shen X, et al.
Journal of Ophthalmology, 2018 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service