Z130931
Nalgene® Griffin beakers
capacity 50 mL, translucent polypropylene
Synonym(s):
PP beaker, nalgene PP beaker
All Photos(1)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Recommended Products
material
translucent polypropylene
feature
graduations
packaging
pkg of 12 ea
manufacturer/tradename
Nalgene 1201-0050
capacity
50 mL
Related Categories
General description
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Other Notes
Browse all plastic beakers
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Aaron F Cipriano et al.
Acta biomaterialia, 62, 397-417 (2017-08-19)
This article reports anodization of Mg in KOH electrolyte and the associated surface, degradation, and biological properties for bioresorbable implant applications. The preparation procedures for electrodes and anodization setup significantly enhanced reproducibility of samples. The results of anodization performed at