Key Documents
THSD1146
ThalesNano MidiCart catalyst cartridge system
L × diam. 116 mm × 9.2 mm, Raney cobalt
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
41103817
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS D1146
L × diam.
116 mm × 9.2 mm
General description
MiDiCarts are suitable for use only with Thalesnano H-Cube MidiCart reactors.
Legal Information
MidiCart is a trademark of ThalesNano Nanotechnology Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)