Skip to Content
Merck
All Photos(1)

Key Documents

SLW1612/03

Pyrex® Vacuum desiccator stopcock, Rotaflo® stopcock

joint: 24/29

Synonym(s):

Pyrex, SciLabware 1612/03

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

Joint

joint: 24/29

packaging

pkg of 1 ea

manufacturer/tradename

1612/03
SciLabware

General description

Stopcock
  • For use with Pyrex vacuum desiccators
  • With Rotaflo stopcock for vacuum tight seal

Legal Information

PYREX is a registered trademark of Corning, Inc.
Rotaflo is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service