SIK7701-10
12-Aminododecylphosphonic acid hydrochloride salt
≥95%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H29ClNO3P
Molecular Weight:
301.79
UNSPSC Code:
12352103
NACRES:
NA.23
Assay
≥95%
form
solid
shelf life
3 years under the recommended conditions
color
white
Related Categories
Application
This coupling agent specifically reacts with cabonyl functions such as aldehydes or ketones to form an imine ligation.
Legal Information
Product of SiKEMIA
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service