GF02174217
Platinum-tungsten alloy, Pt92W8
1m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation
Synonym(s):
PT105825
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
description
polyimide insulation
form
wire (insulated)
manufacturer/tradename
Goodfellow 021-742-17
L
1 m
conductor diam.
0.025 mm
insulation thickness
0.007 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service