Skip to Content
Merck
All Photos(1)

Key Documents

DWK223684

WHEATON® serum vial

tube capacity (3 mL), crimp-top

Synonym(s):

223684

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

flat bottom
transparent borosilicate glass tube

sterility

non-sterile

feature

without cap
: no

packaging

case of 144 ea Shrink-wrapped modules

manufacturer/tradename

WHEATON® 223684

tube capacity

(3 mL)

tube diam.

(17 mm)

fitting

(Pushfit)

Looking for similar products? Visit Product Comparison Guide

General description

Serum tubing vials. Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintains the purity of the contents.
  • Clear vials manufactured from low extractable borosilicate glass that conforms to USP type 1 and ASTM E 438 Type 1, Class A requirements
  • Amber vials converted from Type I borosilicate glass tubing and conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
  • Amber glass conforms to USP 1 requirements to protect light-sensitive samples
  • Tubular design provides excellent clarity and dimensional consistency from vial to vial
  • Specially designed bottom radius adds strength for lyophilization applications
  • Lighter weight compared to molded bottles
  • Shrink-wrapped modules reduce particulate contamination
  • Autoclavable

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service