Key Documents
CAD9915
Animal feeding needle, sterile, disposable, malleable 304 SS
size 15G, L × diam. 3 in. × 2.9 mm , ball
About This Item
UNSPSC Code:
42142609
NACRES:
NB.03
size
15G
packaging
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
3 in. × 2.9 mm , ball
Related Categories
General description
  • Designed to enable the researcher to shape the needle to specific needs
  • Needles come straight and are easily bent to any desired shape
  • Made of type 304 stainless steel tubing
  • Plastic Luer hubs
  • Silicone tips for smoothest possible insertion
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Choose from one of the most recent versions:
Certificates of Analysis (COA)