Skip to Content
Merck
All Photos(1)

Key Documents

C43609

Sigma-Aldrich

4-Chloro-4′-fluorobutyrophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CO(CH2)3Cl
CAS Number:
Molecular Weight:
200.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.5255 (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(cc1)C(=O)CCCCl

InChI

1S/C10H10ClFO/c11-7-1-2-10(13)8-3-5-9(12)6-4-8/h3-6H,1-2,7H2

InChI key

HXAOUYGZEOZTJO-UHFFFAOYSA-N

Physical properties

Material darkens in storage with no loss in purity

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seth Y Ablordeppey et al.
Bioorganic & medicinal chemistry, 16(15), 7291-7301 (2008-07-04)
The synthesis and exploration of novel butyrophenones have led to the identification of a diazepane analogue of haloperidol, 4-[4-(4-chlorophenyl)-1,4-diazepan-1-yl]-1-(4-fluorophenyl)butan-1-one (compound 13) with an interesting multireceptor binding profile. Compound 13 was evaluated for its binding affinities at DA subtype receptors, 5HT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service