Skip to Content
Merck
All Photos(1)

Key Documents

BR759705

BRAND® disposal bag

polypropylene

Synonym(s):

Bags, Disposal Bags

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

material

polypropylene

manufacturer/tradename

BRAND 759705

L × W

300 mm × 200 mm

Looking for similar products? Visit Product Comparison Guide

General description

Clear polypropylene laboratory disposal bags are designed for safe and convenient disposal of laboratory waste. These bags are transparent, allowing for easy identification of contents. They are made from durable polypropylene material, ensuring resistance to punctures and tears, and are suitable for containing a variety of non-sharp laboratory items such as tissues, gloves, and other non-hazardous materials.

Application

Waste disposal

Features and Benefits

  • Made of polypropylene
  • Autoclavable
  • Sharp objects, such as needles, should never be placed in this bag

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service