BR114864
BRAND® storage box
Holds 81 x 3,4 or 5mL tubes
Synonym(s):
Accessories, Cryogenic Tubes, Sample Storage, Storing box, Tubes
All Photos(1)
About This Item
material
polycarbonate
feature
Holds 81 x 3,4 or 5mL tubes
packaging
pack of 5 ea
manufacturer/tradename
BRAND 114864
size
132x132x95 mm
Related Categories
General description
For cryogenic tubes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG