Skip to Content
Merck
All Photos(2)

Key Documents

907243

Sigma-Aldrich

Methyl (4-bromobenzoyl)-L-cysteinate

≥95%

Synonym(s):

Chemoselective thiol probe for detection of electrophilic metabolites, Cysteine thiol probe, Pharmacophore probe for electrophilic natural products

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12BrNO3S
CAS Number:
Molecular Weight:
318.19
UNSPSC Code:
12352101

Assay

≥95%

form

powder

storage temp.

2-8°C

Application

Methyl (4-bromobenzoyl)-L-cysteinate is a chemoselective thiol probe for detection of electrophilic metabolites. As it is often difficult to isolate low-abundance natural products from extracts, metabolities with bioactivity or therapeutic potential may be missed. Castro-Falcón, et al developed a reactivity-guided approach using thiolate probes to discover electrophilic natural products - a cysteine-based probe and thiophenol-based probe. Both consist of
  • A chemoselective reagent for covalent reactivity with a pharmacophore,
  • A conspicuous UV/VIS or mass spectrometric (MS) tag,
  • A linker between the tag and reagent. Methyl (4-bromobenzoyl)-L-cysteinate contains a bromine MS tag and phenyl bromide UV tag that was demonstrated to chemoselectively label enone-, β-lactam-, and β-lactone-based natural products.

Complementarily, a thiophenol probe (cat# 125237) with a chlorine MS tag and phenyl chloride UV tag labeled epoxide-based natural products. These probes can be used in diverse extracts (bacterial, fungal, plant) to chemically faciliate access to high-value metabolites.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Reimer et al.
Journal of natural products, 80(1), 126-133 (2017-01-06)
To date, 16 members of the ammosamide family of natural products have been discovered, and except for ammosamide D each of these metabolites is characterized by an unusual chlorinated pyrrolo[4,3,2-de]quinoline skeleton. Several ammosamides have been shown to inhibit quinone reductase
Gabriel Castro-Falcón et al.
ACS chemical biology, 11(8), 2328-2336 (2016-06-14)
New methods are urgently needed to find novel natural products as structural leads for the development of new drugs against emerging diseases such as cancer and multiresistant bacterial infections. Here we introduce a reactivity-guided drug discovery approach for electrophilic natural
Gabriel Castro-Falcón et al.
Journal of medicinal chemistry (2018-11-02)
Using a novel chemistry-based assay for identifying electrophilic natural products in unprocessed extracts, we identified the PI3-kinase/mTOR dual inhibitor neolymphostin A from Salinispora arenicola CNY-486. The method further showed that the vinylogous ester substituent on the neolymphostin core was the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service