Skip to Content
Merck
All Photos(1)

Documents

743534

Sigma-Aldrich

Phenyl[3-(trifluoromethyl)phenyl]iodonium triflate

≥98% (HPLC)

Synonym(s):

Phenyl[3-(trifluoromethyl)phenyl]iodonium trifluoromethanesulfonate, [3-(Trifluoromethyl)phenyl]phenyliodonium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9F6IO3S
CAS Number:
Molecular Weight:
498.18
Beilstein:
10503310
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98% (HPLC)

form

solid

composition

carbon, 32.9-34.6% CH

reaction suitability

reagent type: oxidant

SMILES string

[O-]S(=O)(=O)C(F)(F)F.FC(F)(F)c1cccc([I+]c2ccccc2)c1

InChI

1S/C13H9F3I.CHF3O3S/c14-13(15,16)10-5-4-8-12(9-10)17-11-6-2-1-3-7-11;2-1(3,4)8(5,6)7/h1-9H;(H,5,6,7)/q+1;/p-1

InChI key

OSSLDNUSYGVAIB-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service