Skip to Content
Merck
All Photos(2)

Key Documents

517585

Sigma-Aldrich

Cadmium acetylacetonate

≥99.9% trace metals basis

Synonym(s):

2,4-Pentanedione cadmium derivative, Cd(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(C5H7O2)2
CAS Number:
Molecular Weight:
310.63
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

core: cadmium
reagent type: catalyst

mp

209-214 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Cd]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Cd/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

JXULEGTWCQDLTM-FDGPNNRMSA-L

General description

Atomic number of base material: 48 Cadmium

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Bone

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongming Zhou et al.
Nature communications, 11(1), 2944-2944 (2020-06-12)
Non-noble metal plasmonic materials, e.g. doped semiconductor nanocrystals, compared to their noble metal counterparts, have shown unique advantages, including broadly tunable plasmon frequency (from visible to infrared) and rich surface chemistry. However, the fate and harvesting of hot electrons from
Katarzyna Ostrowska et al.
Inorganic chemistry, 54(17), 8423-8435 (2015-08-21)
Analysis of the spectral properties and structural differences of two turn-on ratiometric fluorescent receptors for Zn(2+) and Cd(2+) ions, derivatives of pyrrolo[2,3-b]quinoxaline (2), and earlier published 3 (Ostrowska et al. CrystEngComm 2015, 17, 498-502) was performed. Both ligands are E/Z
Zeke Liu et al.
Nature communications, 10(1), 1394-1394 (2019-03-29)
Metal-oxide nanocrystals doped with aliovalent atoms can exhibit tunable infrared localized surface plasmon resonances (LSPRs). Yet, the range of dopant types and concentrations remains limited for many metal-oxide hosts, largely because of the difficulty in establishing reaction kinetics that favors
Taeyjuana Y Lyons et al.
Langmuir : the ACS journal of surfaces and colloids, 33(12), 3018-3027 (2017-03-01)
The increasing commercialization of consumer electronic products that make use of II-VI semiconductor quantum dots (QDs) has raised significant concerns about their impact on natural systems and human health once they are released into the environment at the end of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service