Skip to Content
Merck
All Photos(2)

Key Documents

245852

Sigma-Aldrich

Propyl formate

97%

Synonym(s):

n-Propyl methanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOCH2CH2CH3
CAS Number:
Molecular Weight:
88.11
Beilstein:
1739272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.03 (vs air)

Quality Level

vapor pressure

100 mmHg ( 29.5 °C)
50 mmHg ( 20 °C)

Assay

97%

form

liquid

autoignition temp.

851 °F

refractive index

n20/D 1.375 (lit.)

bp

80-81 °C (lit.)

mp

−93 °C (lit.)

solubility

water: soluble 45 part(lit.)
alcohol: miscible(lit.)
diethyl ether: miscible(lit.)

density

0.904 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)OCCC

InChI

1S/C4H8O2/c1-2-3-6-4-5/h4H,2-3H2,1H3

InChI key

KFNNIILCVOLYIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propyl formate was used in the synthesis of the radicals - CH3CH2CHOCHO, CH3CHCH2OCHO, and CH2CH2CH2OCHO by TiCl3.H2O2 radical-generating method[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

EPR study of radicals derived from n-propyl formate.
Journal of Magnetic Resonance, 39(1), 137-140 (1969)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service