Merck
  • MiR-761 Promotes Progression and Metastasis of Non-Small Cell Lung Cancer by Targeting ING4 and TIMP2.
MiR-761 Promotes Progression and Metastasis of Non-Small Cell Lung Cancer by Targeting ING4 and TIMP2.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-08-19)
An Yan, Chunyu Yang, Zhibo Chen, Chunhong Li, Li Cai
ABSTRACT
The aim of this study was to investigate the role of microRNA miR-761 in the progression and metastasis of non-small cell lung cancer (NSCLC), and the mechanisms by which miR-761 regulates cell proliferation and metastatic activity of NSCLC cell lines. Quantitative real-time PCR (qRT-PCR) was used to assess miR-761 expression in NSCLC serum and tissue. MTT, wound healing, and transwell assays were performed to examine the role of miR-761 in regulation of cell proliferation and metastatic activity in NSCLC cell lines. In addition, the correlations of miR-761 expression with clinical-pathologic factors were statistically analyzed. Finally, we investigated whether miR-761 promotes proliferation and metastasis in NSCLC cell lines by targeting ING4 (inhibitor of growth family, member 4) and TIMP2 (tissue inhibitor of metalloproteinase 2). MiR-761 was significantly upregulated in both NSCLC serum and tissues as compared to normal participants and paired noncancerous tissues respectively. Ectopic expression of miR-761 promoted cell proliferation and metastasis in H460 cells, while miR-761 inhibitor reduced proliferation rates and metastasis in H23 cells. Furthermore, luciferase reporter assay and functional analyses indicated that miR-761 directly targeted ING4 and TIMP2. miR-761 promotes progression and metastasis of NSCLC by targeting ING4 and TIMP2.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ing4
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TIMP2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Timp2