Merck
  • Effects of Diabetes Mellitus on Fibrin Clot Structure and Mechanics in a Model of Acute Neutrophil Extracellular Traps (NETs) Formation.
Effects of Diabetes Mellitus on Fibrin Clot Structure and Mechanics in a Model of Acute Neutrophil Extracellular Traps (NETs) Formation.
International journal of molecular sciences (2020-10-01)
Judith J de Vries, Tamara Hoppenbrouwers, Cristina Martinez-Torres, Rezin Majied, Behiye Özcan, Mandy van Hoek, Frank W G Leebeek, Dingeman C Rijken, Gijsje H Koenderink, Moniek P M de Maat
ABSTRACT
Subjects with diabetes mellitus (DM) have an increased risk of arterial thrombosis, to which changes in clot structure and mechanics may contribute. Another contributing factor might be an increased formation of neutrophil extracellular traps (NETs) in DM. NETs are mainly formed during the acute phase of disease and form a network within the fibrin matrix, thereby influencing clot properties. Previous research has shown separate effects of NETs and DM on clot properties, therefore our aim was to study how DM affects clot properties in a model resembling an acute phase of disease with NETs formation. Clots were prepared from citrated plasma from subjects with and without DM with the addition of NETs, induced in neutrophils by S. aureus bacteria or phorbol myristate acetate (PMA). Structural parameters were measured using scanning electron microscopy, mechanical properties using rheology, and sensitivity to lysis using a fluorescence-based fibrinolysis assay. Plasma clots from subjects with DM had significantly thicker fibers and fewer pores and branch points than clots from subjects without DM. In addition, fibrinolysis was significantly slower, while mechanical properties were similar between both groups. In conclusion, in a model of acute NETs formation, DM plasma shows prothrombotic effects on fibrin clots.
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
Pricing and availability is not currently available.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Pricing and availability is not currently available.