Skip to Content
Merck
All Photos(1)

Key Documents

V900652

Sigma-Aldrich

DL-Isoleucine

Vetec, reagent grade, 98%

Synonym(s):

DL-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721790
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

290 °C (dec.) (lit.)

SMILES string

CC[C@@H](C)[C@@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m1/s1

InChI key

AGPKZVBTJJNPAG-RFZPGFLSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A mixture of 4 stereoisomers.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service