V900192
Sodium tetrafluoroborate
Vetec™, reagent grade, 98%
Synonym(s):
Sodium fluoroborate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
NaBF4
CAS Number:
Molecular Weight:
109.79
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
reagent grade
form:
powder or crystals
grade
reagent grade
product line
Vetec™
Assay
98%
form
powder or crystals
density
2.47 g/mL at 25 °C (lit.)
SMILES string
[Na+].F[B-](F)(F)F
InChI
1S/BF4.Na/c2-1(3,4)5;/q-1;+1
InChI key
KGJZTOFHXCFQIV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service