Skip to Content
Merck
All Photos(1)

Documents

V800258

Sigma-Aldrich

Methanol

≥99.5%, Analytical Reagent grade, Vetec

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
15101803
PubChem Substance ID:

product name

Methanol, AR, ≥99.5%

grade

AR

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kentaro Fujiwara et al.
EJNMMI research, 4, 29-29 (2014-07-10)
ROBO1 is a membrane protein that functions in axon guidance. ROBO1 contributes to tumour metastasis and angiogenesis and may have potential as a target protein of immunotherapy because ROBO1 is specifically expressed at high levels in hepatocellular carcinoma. In this
Haruka Oda et al.
Biomaterials, 56, 86-91 (2015-05-03)
A large number of lineage-committed progenitor cells are required for advanced regenerative medicine based on cell engineering. Due to their ability to differentiate into multiple cells lines, multipotent stem cells have emerged as a vital source for generating transplantable cells
Timo Heidt et al.
Circulation research, 115(2), 284-295 (2014-05-03)
Macrophages populate the steady-state myocardium. Previously, all macrophages were thought to arise from monocytes; however, it emerged that, in several organs, tissue-resident macrophages may self-maintain through local proliferation. Our aim was to study the contribution of monocytes to cardiac-resident macrophages
Nicolas Vodovar et al.
European heart journal, 35(48), 3434-3441 (2014-08-27)
Increases in plasma B-type natriuretic peptide (BNP) concentrations in those with acutely decompensated heart failure (ADHF) has been mainly attributed to an increase in NPPB gene transcription. Recently, proBNP glycosylation has emerged as a potential regulatory mechanism in the production
Panagiotis Ntziachristos et al.
Nature, 514(7523), 513-517 (2014-08-19)
T-cell acute lymphoblastic leukaemia (T-ALL) is a haematological malignancy with a dismal overall prognosis, including a relapse rate of up to 25%, mainly because of the lack of non-cytotoxic targeted therapy options. Drugs that target the function of key epigenetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service