Skip to Content
Merck
All Photos(1)

Key Documents

V000645

Sigma-Aldrich

Iodomethane

for synthesis, 99.0%

Synonym(s):

Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Beilstein:
969135
EC Number:
MDL number:
PubChem Substance ID:

grade

for synthesis

Quality Level

vapor density

4.89 (vs air)

vapor pressure

24.09 psi ( 55 °C)
7.89 psi ( 20 °C)

Assay

99.0%

refractive index

n20/D 1.531 (lit.)

bp

41-43 °C (lit.)

mp

−64 (lit.)

density

2.28 g/mL at 25 °C (lit.)

SMILES string

CI

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.8 °F - closed cup

Flash Point(C)

32.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riet Bammens et al.
Glycobiology, 25(6), 669-682 (2015-01-23)
The congenital disorders of glycosylation (CDG), a group of inherited diseases characterized by aberrant glycosylation, encompass a wide range of defects, including glycosyltransferases, glycosidases, nucleotide-sugar transporters as well as proteins involved in maintaining Golgi architecture, pH and vesicular trafficking. Mutations
Ann-Charlotte Almstrand et al.
Analytical and bioanalytical chemistry, 407(17), 5045-5052 (2015-03-05)
Lipidomic analysis of the complex mixture of lipids isolated from biological systems can be a challenging process that often involves tandem mass spectrometry and interpretation of both precursor ions and product ions relative to the molecular structure of the lipids.
Joel E Schmidt et al.
Angewandte Chemie (International ed. in English), 53(32), 8372-8374 (2014-06-26)
Crystalline molecular sieves are used in numerous applications, where the properties exploited for each technology are the direct consequence of structural features. New materials are typically discovered by trial and error, and in many cases, organic structure-directing agents (OSDAs) are
Hagay Kohay et al.
Environmental science & technology, 49(15), 9280-9288 (2015-07-01)
The presence of nanoconcentrations of persistent pharmaceuticals in treated wastewater effluent and in surface water has been frequently reported. A novel organic-inorganic hybrid sorbent based on adsorbing quarternized poly vinylpyridinium-co-styrene (QPVPcS) to montmorillonite (MMT) was designed for the removal of
Xiangfeng Meng et al.
Applied microbiology and biotechnology, 99(14), 5885-5894 (2015-01-15)
Glucansucrases are exclusively found in lactic acid bacteria and synthesize a variety of α-glucans from sucrose. They are large multidomain enzymes belonging to the CAZy family 70 of glycoside hydrolase enzymes (GH70). The crystal structure of the N-terminal truncated GTF180

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service