Skip to Content
Merck
All Photos(1)

Key Documents

SAAR191235

Silver nitrate solution

0.1 N (0.1 mol/L)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302

packaging

glass bottle of 4 × 2.5 L (SAAR191235LC)

concentration

0.1 N (0.1 mol/L)

storage temp.

2-30°C

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service