PHR9174
Ethyl 2-aminobenzoate
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Benzocaine impurity D (PhEur), Ethyl anthranilate
Select a Size
€304.00
Select a Size
About This Item
€304.00
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
vapor density
5.7 (vs air)
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
refractive index
n20/D 1.564 (lit.)
bp
129-130 °C/9 mmHg (lit.)
mp
13-15 °C (lit.)
density
1.117 g/mL at 25 °C (lit.)
application(s)
pharmaceutical small molecule
storage temp.
2-8°C
SMILES string
CCOC(=O)c1ccccc1N
InChI
1S/C9H11NO2/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6H,2,10H2,1H3
InChI key
TWLLPUMZVVGILS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Application
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service