Skip to Content
Merck
All Photos(1)

Documents

PHR9150

Supelco

Paclitaxel impurity K

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

7-O-(Triethylsilanyl)paclitaxel, 7-O-(Triethylsilyl)taxol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H65NO14Si
CAS Number:
Molecular Weight:
968.17
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

API family

paclitaxel

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

InChI

1S/C53H65NO14Si/c1-10-69(11-2,12-3)68-38-28-39-52(30-63-39,67-33(6)56)44-46(66-48(60)36-26-20-15-21-27-36)53(62)29-37(31(4)40(50(53,7)8)43(64-32(5)55)45(58)51(38,44)9)65-49(61)42(57)41(34-22-16-13-17-23-34)54-47(59)35-24-18-14-19-25-35/h13-27,37-39,41-44,46,57,62H,10-12,28-30H2,1-9H3,(H,54,59)/t37-,38-,39+,41-,42+,43+,44-,46-,51+,52-,53+/m0/s1

InChI key

LKHIBBPOJARUFY-RGZLSWIJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download
your certificate at: http://www.sigma-aldrich.com

Application

These Reference Materials are suitable for use in several analytical applications including, but not limited to, method development for qualitative and quantitative analyses, daily calibration, and routine quality control testing.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service