Skip to Content
Merck
All Photos(1)

Documents

PHL84768

Curzerene

phyproof® Reference Substance

Synonym(s):

Isofuranogermacrene, Isogermafurene, Neocurzerene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O
CAS Number:
Molecular Weight:
216.32
UNSPSC Code:
41116107

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

form

liquid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service