Skip to Content
Merck
All Photos(1)

Documents

PHL82650

Senegenin

phyproof® Reference Substance

Synonym(s):

12-(Chloromethyl)-2,3-dihydroxy-27-norolean-13-ene-23,28-dioic acid, Tenuigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H45ClO6
CAS Number:
Molecular Weight:
537.13
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

SMILES string

[H][C@]12[C@](CCC3=C2[C@@H](CCl)C[C@@]4([H])[C@@]3(C)CC[C@]5([H])[C@]4(C)C[C@H](O)[C@H](O)[C@]5(C(O)=O)C)(C(O)=O)CCC(C)(C)C1

InChI

1S/C30H45ClO6/c1-26(2)10-11-30(25(36)37)9-6-17-22(18(30)13-26)16(15-31)12-21-27(17,3)8-7-20-28(21,4)14-19(32)23(33)29(20,5)24(34)35/h16,18-21,23,32-33H,6-15H2,1-5H3,(H,34,35)(H,36,37)/t16-,18+,19+,20-,21+,23+,27+,28+,29+,30-/m1/s1

InChI key

CWHJIJJSDGEHNS-MYLFLSLOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Senegenin (Tenuigenin) is a natural product from Polygala tenuifolia used in Chinese medicine with antioxidative and antiinflammatory activity. Senegenin has been studied for nootropic ability to improve cognition. In several independent studies, Tenuigenin attenuated α-synuclein-induced cytotoxicity and α-synuclein phosphorylation by down-regulating polo-like kinase 3 (PLK3) in α-synuclein cell model of Parkinson′s Disease, improved behavioral Y-maze learning by enhancing synaptic plasticity in mice, and attenuated hepatic ischemia-reperfusion induced cognitive dysfunction by increasing hippocampal N-methyl-D-aspartate (NMDA) receptor NR2B subunit expression in rats.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service