Recommended Products
availability
available only in EU
parameter
200 mL/min flow rate (calibrated)
size
20 × 10 × 10
application(s)
environmental
industrial hygiene
compatibility
for sampling dynamic air flow
General description
This pump is calibrated to draw air at a flow rate of 200 mL/min. This is necessary for dynamic air sampling.
Coupled with the dynamic sampling module (PAMEX001-1EA), it creates a controlled air flow around the dynamic sampling sensors, thus accelerating the capture process of the material.
Coupled with the dynamic sampling module (PAMEX001-1EA), it creates a controlled air flow around the dynamic sampling sensors, thus accelerating the capture process of the material.
Suitability
Fits the following instruments:
PAKSF421
PAMEX001
PAKSF421
PAMEX001
Other Notes
Weight: 0.8
Temperature conditions: Room temperature
Temperature conditions: Room temperature
Legal Information
Profil'Air is a registered trademark of Ethera S.A.
Choose from one of the most recent versions:
Certificates of Analysis (COA)