G1007465
HPLC Analysis of Alfuzosin and Related Substances on Chromolith® HighResolution RP-18 endcapped
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
sample preparation: Dissolve 40.0 mg of the sample in 100 mL of mobile phase. 0.4 mg/mL (400 ppm) of USP Alfuzosin System Suitability Mixture RS in Mobile phase
column: Chromolith® HighResolution RP-18 endcapped, 150 x 4.6 mm (1.52023.0001)
mobile phase: [A] 5 ml of perchloric acid in 900 mL Milli-Q water. Adjust pH to 3.5 with 2M NaOH. Diluter to 1000 mL with water; [B] acetonitrile; [C] THF; (80:20:1, A:B:C)
flow rate: 1 mL/min
pressure: 994 psi (69 Bar)
column temp.: 25 °C
detector: 254 nm cell: 10 μL
injection: 10 μL
sample: 0.40 μg/mL of Alfuzosin Hydrochloride in Mobile phase from the Sample solution
suitability
application for HPLC
application(s)
pharmaceutical (small molecule)
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service