G005714
HPLC Analysis of Organic Acids on Ascentis® Express RP-Amide
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase: [A] 0.1% (v/v) TFA in [B] water
flow rate: 0.3 mL/min
column temp.: 35 °C
detector: UV, 210 nm
injection: 1 μL
sample: 0.2 - 4 g/L in mobile phase
suitability
application for HPLC
application(s)
chemicals and industrial polymers
food and beverages
vitamins, nutraceuticals, and natural products
Analysis Note
Legal Information
Analyte
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service