G005379
HPLC Analysis of Basic Peptides (Angiotensin Analogs) on BIOshell A160 Peptide C18
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: BIOshell A160 Peptide C18, 10 cm x 3 mm I.D., 2.7 μm particles (66908-U)
mobile phase: [A] 0.1% (v/v) formic acid, pH 4.0 (titrated with ammonium hydroxide); [B] 50:50, (0.175% formic acid in water, pH 4.0) : acetonitrile
gradient: 20 to 50% B in 15 min
flow rate: 0.6 mL/min
pressure: 3002 psi (207 bar)
column temp.: 35 °C
detector: UV, 215 nm
injection: 2.0 μL
sample: 0.5 g/L ea. peptide
suitability
application for HPLC
application(s)
life science and biopharma
pharmaceutical (small molecule)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service