AmberTec™ UP550 Ion Exchange Resin
hydroxide form, 16-50 mesh, anionic
Synonym(s):
strong anion exchange resin
About This Item
Recommended Products
product name
AmberTec™ UP550 OH Anion Exchange Resin,
description
Trimethylamine functionalised copolymer of styrene and divinylbenzene in the hydroxide form (CAS No. 69011-18-3; Water (CAS No. 7732-18-5)
formerly Dowex Monosphere 550A UPW OH
form
beads
packaging
pkg of 1 kg (14016-U)
pkg of 100 g (14015-U)
pkg of 20 kg (14019-U)
pkg of 5 kg (14018-U)
manufacturer/tradename
Du Pont AmberTec™
parameter
60 °C max. temp.
technique(s)
LPLC: suitable
matrix
styrene-divinylbenzene
matrix active group
quaternary amine functional group
particle size
590 μm std dev ±50
operating pH
0-14
capacity
1.0 eq/L by wetted bed volume
55-65 % water retention capacity ((OH))
separation technique
anion exchange
Related Categories
Application
Footnote
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service