Skip to Content
Merck
All Photos(1)

Documents

819008

TSKgel® Octadecyl-4PW (7 µm) HPLC Columns

L x I.D. 1.5 cm × 3.2 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Guard Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® Octadecyl-4PW Guard Cartridge, phase C18 (octadecyl), L × I.D. 1.5 cm × 3.2 mm, 7 μm particle size, pkg of 3 ea

material

stainless steel

product line

TSKgel®

packaging

pkg of 3 ea

manufacturer/tradename

Tosoh Octadecyl-4PW, 19008

extent of labeling

1 meq/mL ligand density (estimate)

parameter

no limitation salt concentration
0-100% organic solvent
0.22 mL/min flow rate
5-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1.5 cm × 3.2 mm

matrix

polymer particle platform

matrix active group

C18 (octadecyl) phase

particle size

7 μm

pore size

500 Å

operating pH

2-12

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Highly cross-linked polymethacrylate base material of TSKgel Octadecyl-4PW provides excellent stability in high pH buffer systems and can withstand rigorous cleaning with either acid or base. The 500Ã… pore allows for anaylsis of proteins up to 200kDa while the particle size offerings allow for analytical, semi-preparative and preparative scale separations.
TSKgel Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No. 819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No. 819018.

Reversed-phase liquid chromatography (RPLC) is the preferred method for analysis and purification of polar and nonpolar compounds that are soluble in mixtures of water and organic solvents.

Physical form

Shipped in 60% methanol - 40% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service