Skip to Content
Merck
All Photos(1)

Documents

808639

TSKgel® Sugar AXI (8 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, trimethylamine phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® Sugar AXI HPLC Column, phase trimethylamine, L × I.D. 15 cm × 4.6 mm, 8 μm particle size

material

stainless steel

product line

TSKgel®

feature

counter ion HBO3-

manufacturer/tradename

Tosoh 08639

parameter

≤20% organic solvent
0.2-0.4 mL/min flow rate
25-80 °C temperature
30 bar max. pressure (60-80 °C)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

polystyrene-divinylbenzene

matrix active group

trimethylamine phase

particle size

8 μm

operating pH

7-10

separation technique

anion exchange

General description

TSKgel® Sugar AXI columns are packed with 8μm porous spherical PS/DVB particles which are modified with a strong anion exchanger. They are used for the isocratic separation and analysis of monosaccharides, disaccharides, and sugar alcohols.For gradient separation of carbohydrates where higher back pressures may be generated, please see the TSKgel Sugar AXG column, PN 808640, which is packed with 10μm particles.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service