Key Documents
64716-U
Supelco
Screw Top Mini Flask with Hole Cap and Septum
volume 25 mL, pkg of 12 ea
About This Item
UNSPSC Code:
41115724
NACRES:
NB.21
material
clear borosilicate glass flask
packaging
pkg of 12 ea
volume
25 mL
Features and Benefits
Heat resistant borosilicate glass flasks provide an inert seal when used with a hole cap and PTFE silicone septum (included) or a Mininert valve. Excellent for liquid-liquid extraction of trihalomethanes from drinking water. Also useful for sample collection, solution preparations, derivatization reactions, extractions, and many other applications.
Other Notes
Replacement cap for the Mini Flask is part number 27176, the replacement septa is part number 27177.
Choose from one of the most recent versions:
Certificates of Analysis (COA)