Skip to Content
Merck
All Photos(1)

Key Documents

581383-U

Supelco

BIOshell IgG 1000 Å C18 (2.7 μm) HPLC Columns

L × I.D. 5 cm × 1 mm, HPLC Column

Synonym(s):

TMBIOSHELLIGG1000C1827HPLCCOLUMN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

BIOshell IgG 1000 Å C18, 2.7 μm HPLC Column, column L × I.D. 5 cm × 1 mm

material

stainless steel hardware

Quality Level

Agency

suitable for USP L1

product line

BIOshell

feature

endcapped

packaging

pkg of 1 unit

extent of labeling

1.4% Carbon loading

technique(s)

HPLC: suitable

column L × I.D.

5 cm × 1 mm

surface area

22 m2/g

matrix

Fused-Core® particle platform

matrix active group

C18 (diisobutyloctadecylsilane) bonding phase

particle size

2.7 μm

pore size

1000 Å pore size

operating pH range

1-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell IgG 1000 Å C18 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1000 Å) Fused-Core® particle design. The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The sterically protected C18 bonded phase of BIOshell IgG 1000 Å C18 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.

Analysis Note

The target analytes include monoclonal antibodies, antibody-drug conjugates, antibody fragments and large proteins with molecular weights up to 500 kDa.

Legal Information

Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service