56058-U
Omnifit® Tube Endfittings
for use with 1/16 in. O.D. tubing, black polypropylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
SB.52
Recommended Products
material
black polypropylene
packaging
pkg of 10 ea
technique(s)
LPLC: suitable
fitting
1/4-28 male UNF nut (Gripper Fitting required)
compatibility
for use with 1/16 in. O.D. tubing
Looking for similar products? Visit Product Comparison Guide
General description
Each endfitting has an internal taper to hold the tubing centrally; includes a lock nut for panel mounting.
Gripper fitting required (purchase separately):
for 1/16in. O.D. tubing p/n 57417
for 1/8in. O.D. tubing p/n 57418
Gripper fitting required (purchase separately):
for 1/16in. O.D. tubing p/n 57417
for 1/8in. O.D. tubing p/n 57418
Legal Information
Omnifit is a registered trademark of Diba Industries
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Study of potential leakage on several stressed fittings for hydrogen pressures up to 700 bar.
Deborah, H. D.
International Journal of Hydrogen Energy, 37(22), 17509-17516 (2012)
Salvatore, F.
Liquid Chromatography: Fundamentals and Instrumentation (2013)
John W.D.
Troubleshooting LC Systems (1989)
Liquid chromatography on an amide stationary phase with post-column derivatization and fluorimetric detection for the determination of streptomycin and dihydrostreptomycin in foods.
Pilar, V. N.
Talanta, 72(2), 808-812 (2007)
Interpreting the difference between conventional and bi-directional plate-height measurements in liquid chromatography
Hui, G. D.
Journal of Chromatography A, 1217(40), 6214-6229 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service