Skip to Content
Merck
All Photos(1)

Documents

51851

Supelco

Diethylenetriamine

analytical standard

Synonym(s):

2,2′-Diaminodiethylamine, 2,2′-Iminodiethylamine, Bis(2-aminoethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2)2NH
CAS Number:
Molecular Weight:
103.17
Beilstein:
605314
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

grade

analytical standard

Quality Level

vapor density

3.6 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥98.5% (GC)

autoignition temp.

676 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.7 %

refractive index

n20/D 1.483-1.485
n20/D 1.484 (lit.)

bp

199-209 °C (lit.)

mp

−35 °C (lit.)

density

0.955 g/mL at 25 °C (lit.)

SMILES string

NCCNCCN

InChI

1S/C4H13N3/c5-1-3-7-4-2-6/h7H,1-6H2

InChI key

RPNUMPOLZDHAAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service