Documents
507601
Supelco
Vial, Chromacol
volume 2 mL, clear glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mL/ft, thread for 8-425, Chromacol 2-SV, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
material
clear glass vial
feature
closure type screw top vial
vial type screw top
packaging
pkg of 100 ea
manufacturer/tradename
Chromacol 2-SV
O.D. × H × I.D.
12 mm × 32 mm × 4.6 mL/ft
volume
2 mL
fitting
thread for 8-425
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)